| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200029985.mm.1 |
NM_001141949.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200029985.mm.1 |
NM_001141949.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200029985.mm.1 |
TC0200003528.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200029986.mm.1 |
NM_001141948.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200029986.mm.1 |
NM_001141948.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200029986.mm.1 |
NM_001141949.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200029986.mm.1 |
NM_001141949.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200029986.mm.1 |
TC0200003528.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200029988.mm.1 |
NM_001141948.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200029988.mm.1 |
NM_001141948.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200029988.mm.1 |
NM_001141949.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200029988.mm.1 |
NM_001141949.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200029988.mm.1 |
NM_019401.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200029988.mm.1 |
NM_019401.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200029988.mm.1 |
TC0200003528.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200029991.mm.1 |
NM_001141948.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200029991.mm.1 |
NM_001141948.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200029991.mm.1 |
NM_001141949.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200029991.mm.1 |
NM_001141949.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200029991.mm.1 |
NM_019401.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200029991.mm.1 |
NM_019401.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200029991.mm.1 |
TC0200003528.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200029984.mm.1 |
NM_001141948.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200029984.mm.1 |
NM_001141948.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200029984.mm.1 |
NM_001141949.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200029984.mm.1 |
NM_001141949.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200029984.mm.1 |
NM_019401.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200029984.mm.1 |
NM_019401.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200029984.mm.1 |
TC0200003528.mm.1 |
MTA TC |