Assay Details
| Gene Symbol: Hoxa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008265.3 | NP_032291.1 | 1 - 2 | 521 | 68 | |
| GenBank mRNA | AK076098.1 | - | 1 - 2 | 522 | 68 | |
| BC107172.2 | - | 2 - 3 | 228 | 40044111 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
multicellular organism development
anterior/posterior pattern specification
embryonic skeletal system morphogenesis
DNA binding
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding
transcription, DNA-templated
regulation of transcription, DNA-templated
multicellular organism development
anterior/posterior pattern specification
embryonic skeletal system morphogenesis
DNA binding
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24448936 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24448934 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24448932 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24448935 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24448933 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2219982 | NM_008265.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2219982 | AK076098.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2219982 | BC107172.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2219982 | NM_008265.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2219982 | AK076098.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2219982 | BC107172.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P443588 | X13538.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P443588 | NM_008265.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P443588 | AK076098.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P638382 | X13538.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P638382 | NM_008265.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P638382 | AK076098.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P638382 | BC107172.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P638382 | M13813.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24448928 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24449006 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24449004 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24448950 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24448924 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24448995 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24448973 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24448957 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24448944 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24448941 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24448937 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24448971 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24448919 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24448940 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24448943 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24448954 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24448996 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24448997 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24448968 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24448961 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24449002 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24448978 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24448965 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24448976 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24448929 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24448975 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24448966 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24448964 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24448932 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24448979 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24448953 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24448927 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24448948 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24448959 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24448970 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24448938 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24449008 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24448946 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24448993 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24448936 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24448977 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24448972 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24448930 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24448998 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24448949 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24448921 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24448958 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24448994 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24448923 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24448951 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24448925 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24449000 | - | - |
| Whole Mouse Genome G4122A | A_52_P443588 | X13538.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P443588 | NM_008265.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P443588 | AK076098.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P638382 | X13538.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P638382 | NM_008265.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P638382 | AK076098.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P638382 | BC107172.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P638382 | M13813.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106188 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05583115 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05299884 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05855216 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24448933 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24448932 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24448935 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24448934 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24448936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24448968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24449008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24448941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24448977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24448996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24448957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24448936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24448932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24448961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24448985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24448953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24448993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24449006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24448964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24448965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24448954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24448970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24449011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24448998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24448943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24448928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24449012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24448929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24448951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24448972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24448986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24448994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24449002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24448966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24448930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24448982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24448976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24448938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24449016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24448921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24448950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24448984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24448980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24449000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24448946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24448923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24448927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24448989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24449014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24448990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24448925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24448978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24448975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24448940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24448949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24448919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24448973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24448987 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24448949 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24449002 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24448936 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24448986 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24448990 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24448973 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24448961 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24448934 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24448928 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24448970 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24448988 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24448946 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24448943 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24448996 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24448940 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24448929 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24448954 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24448964 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24448921 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05517886 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05686418 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24448923 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24448958 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24448968 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24448993 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24448932 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24448980 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24448965 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24448989 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24448966 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05686420 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05517843 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05306838 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24448975 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24448985 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05299888 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24448941 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24449006 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05686431 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24448925 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05517862 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24448950 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24448971 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24448984 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24448927 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24448977 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24448998 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24448957 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05517875 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05686427 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24448938 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24448930 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24449008 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24448976 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24448919 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24448953 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10544765 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10544767 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10544764 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10544766 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420226_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427354_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10544763 | - | - |
| Mouse Genome 430 2.0 Array | 1420227_at | - | - |
| Mouse Genome 430 2.0 Array | 1427354_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1420226_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1420226_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1427354_at | - | - |
| Mouse Genome 430 2.0 Array | 1420227_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 15401_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 15401_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600017842.mm.1 | NM_008265.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600017842.mm.1 | NM_008265.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600017842.mm.1 | TC0600002278.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600017844.mm.1 | NM_008265.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600017844.mm.1 | NM_008265.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600017844.mm.1 | TC0600002278.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600017840.mm.1 | NM_008265.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600017840.mm.1 | NM_008265.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600017840.mm.1 | TC0600002278.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600017841.mm.1 | NM_008265.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600017841.mm.1 | NM_008265.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600017841.mm.1 | TC0600002278.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600017843.mm.1 | NM_008265.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600017843.mm.1 | NM_008265.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600017843.mm.1 | TC0600002278.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1427354_PM_at | - | - |
| Mouse Expression Set 430 | 1427354_at | - | - |
| Murine Genome U74v2 Set | 162402_PM_r_at | - | - |
| Murine Genome U74v2 Set | 97745_at | - | - |
| Murine Genome U74v2 Set | 97745_PM_at | - | - |
| Murine Genome U74v2 Set | 162402_r_at | - | - |
| Murine Genome U74v2 Set | 97747_r_at | - | - |
| Murine Genome U74v2 Set | 97746_PM_f_at | - | - |
| Murine Genome U74v2 Set | 97747_PM_r_at | - | - |
| Murine Genome U74v2 Set | 97746_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427354_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427354_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420226_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427354_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420226_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427354_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 940152 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 940152 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000940152 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2755860 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2755860 | - | - |
| mouse-6 v1.1 (Illumina) | scl29005.2_93-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 940152 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl29005.2_93-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 940152 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2755860 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2755860 | - | - |

Hoxa4
Genomic Map