Assay Details
| Gene Symbol: BC051665 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199148.2 | NP_954599.2 | 3 - 4 | 340 | 84 | |
| GenBank mRNA | AK145559.1 | - | 3 - 4 | 338 | 84 | |
| BC051665.1 | - | 3 - 4 | 325 | 30027545 | 84 | |
More Information
Set Membership: |
|

BC051665
Genomic Map