Assay Details
Gene Symbol: Nhlrc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175340.4 | NP_780549.1 | 1 | 2105 | 143 | |
GenBank mRNA | AK045746.1 | - | 1 | 2076 | 143 | |
BC062644.1 | - | 1 | 2074 | 143 | ||
BX514773.1 | - | 1 | 248 | 143 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
