Assay Details
| Gene Symbol: Fcnaos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK039020.1 | - | 2 - 3 | 775 | 72 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transmembrane protein 141 | Tmem141 |
1110065P14Rik, AI663999, D2Ertd217e |
Anti-sense | Chr. 2 - 25620066 - 25622039 on NCBI Build GRCm38 | |
| Downstream |
predicted gene, 35630 |
Gm35630 | 10.2kb | Sense | Chr. 2 - 25635518 - 25640293 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK039020.1 transcripts.
Set Membership: |
|

Fcnaos
Genomic Map