Assay Details
| Gene Symbol: Arrdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162485.1 | NP_001155957.1 | 2 - 3 | 371 | 93 | |
| NM_178408.3 | NP_848495.2 | 2 - 3 | 371 | 93 | ||
| NR_027876.1 | 2 - 3 | 371 | 93 | |||
| GenBank mRNA | AK088008.1 | - | 2 - 3 | 236 | 93 | |
| AK170018.1 | - | 2 - 3 | 245 | 93 | ||
| BC004091.1 | - | 2 - 3 | 266 | 93 | ||
| BC018501.1 | - | 2 - 3 | 253 | 3662534 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arrdc1
Genomic Map