| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200012317.mm.1 |
NM_207206.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200012317.mm.1 |
NM_207206.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200012317.mm.1 |
TC0200001662.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200012315.mm.1 |
NM_207206.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200012315.mm.1 |
NM_207206.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200012315.mm.1 |
TC0200001662.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200012318.mm.1 |
NM_207206.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200012318.mm.1 |
NM_207206.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200012318.mm.1 |
TC0200001662.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200012316.mm.1 |
NM_207206.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200012316.mm.1 |
NM_207206.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200012316.mm.1 |
TC0200001662.mm.1 |
MTA TC |