Assay Details
| Gene Symbol: AI182371 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243102.1 | NP_001230031.1 | 6 - 7 | 729 | 121 | |
| NM_001243103.1 | NP_001230032.1 | 6 - 7 | 726 | 121 | ||
| NM_178885.4 | NP_849216.2 | 6 - 7 | 726 | 121 | ||
| GenBank mRNA | AK050126.1 | - | 6 - 7 | 730 | 121 | |
| AK050413.1 | - | 6 - 7 | 701 | 121 | ||
| BC019541.1 | - | 6 - 7 | 684 | 121 | ||
| BC037995.1 | - | 6 - 7 | 706 | 121 | ||
| BC094504.1 | - | 6 - 7 | 714 | 5100182 | 121 | |
| BC115979.1 | - | 6 - 7 | 701 | 40104352 | 121 | |
| BC115986.1 | - | 6 - 7 | 701 | 40104349 | 121 | |
More Information
Set Membership: |
|

AI182371
Genomic Map