Assay Details
| Gene Symbol: Lhx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083126.1 | NP_001076595.1 | 7 - 8 | 1056 | 59 | |
| NM_008500.2 | NP_032526.2 | 8 - 9 | 1246 | 59 | ||
| GenBank mRNA | AB031040.1 | - | 7 - 8 | 1056 | 59 | |
| BC065077.1 | - | 7 - 8 | 990 | 6816328 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lhx6
Genomic Map