Assay Details
| Gene Symbol: Ak8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033874.2 | NP_001029046.2 | 1 - 2 | 111 | 89 | |
| GenBank mRNA | AK004439.1 | - | 1 - 2 | 109 | 89 | |
| BC100481.1 | - | 1 - 2 | 108 | 30917849 | 89 | |
| BC109352.2 | - | 1 - 2 | 98 | 40057006 | 89 | |
| BY100469.1 | - | 1 - 2 | 112 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ak8
Genomic Map