Assay Details
| Gene Symbol: Kcnab3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010599.4 | NP_034729.3 | 1 - 2 | 667 | 107 | |
| GenBank mRNA | AK031806.1 | - | 1 - 2 | 648 | 107 | |
| AK044102.1 | - | 1 - 2 | 622 | 107 | ||
| AK046194.1 | - | 1 - 2 | 657 | 107 | ||
| AK049316.1 | - | 1 - 2 | 668 | 107 | ||
| AK083134.1 | - | 1 - 2 | 669 | 107 | ||
| BC017518.1 | - | 1 - 2 | 593 | 4507029 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kcnab3
Genomic Map