Assay Details
| Gene Symbol: Tmem150b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142792.1 | NP_001136264.1 | 4 - 5 | 432 | 68 | |
| GenBank mRNA | BC022651.1 | - | 4 - 5 | 432 | 4216812 | 68 |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem150b
Genomic Map