Assay Details
| Gene Symbol: Il11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290423.1 | NP_001277352.1 | 2 - 3 | 239 | 73 | |
| NM_008350.4 | NP_032376.1 | 2 - 3 | 338 | 73 | ||
| GenBank mRNA | AK132897.1 | - | 2 - 3 | 240 | 73 | |
| BC134354.1 | - | 2 - 3 | 200 | 40054168 | 73 | |
| U03421.1 | - | 2 - 3 | 231 | 73 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001290423.1 transcripts.
Set Membership: |
|

Il11
Genomic Map