Assay Details
| Gene Symbol: Il11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290423.1 | NP_001277352.1 | 3 - 4 | 322 | 80 | |
| NM_008350.4 | NP_032376.1 | 3 - 4 | 421 | 80 | ||
| GenBank mRNA | AK132897.1 | - | 3 - 4 | 323 | 80 | |
| BC134354.1 | - | 3 - 4 | 283 | 40054168 | 80 | |
| U03421.1 | - | 3 - 4 | 314 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Il11
Genomic Map