Assay Details
| Gene Symbol: Ncr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010746.3 | NP_034876.2 | 4 - 5 | 657 | 57 | |
| GenBank mRNA | AJ223765.1 | - | 4 - 5 | 646 | 57 | |
| AK079401.1 | - | 4 - 5 | 658 | 57 | ||
| AK172251.1 | - | 3 - 4 | 690 | 57 | ||
| BC042788.1 | - | 4 - 5 | 627 | 57 | ||
| BC115364.2 | - | 4 - 5 | 658 | 40045898 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P500882 | BC115364.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P500882 | AJ223765.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P500882 | AK079401.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P500882 | AK172251.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P500882 | BC042788.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P500882 | BC115364.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P500882 | AJ223765.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P500882 | AK079401.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P500882 | AK172251.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P500882 | BC042788.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 115813 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931307 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 775841 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 684635 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 197393 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 131650 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P140067 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P500882 | BC115364.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P500882 | AJ223765.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P500882 | AK079401.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P500882 | AK172251.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P500882 | BC042788.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P500882 | BC115364.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P500882 | AJ223765.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P500882 | AK079401.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P500882 | AK172251.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P500882 | BC042788.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P500882 | BC115364.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P500882 | AJ223765.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P500882 | AK079401.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P500882 | AK172251.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P500882 | BC042788.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P500882 | BC115364.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P500882 | AJ223765.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P500882 | AK079401.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P500882 | AK172251.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P500882 | BC042788.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948522 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948524 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948528 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948530 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948527 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948518 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948516 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948515 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948525 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948523 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P140067 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01475357 | - | - |
| Whole Mouse Genome G4122A | A_51_P500882 | BC115364.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P500882 | AJ223765.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P500882 | AK079401.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P500882 | AK172251.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P500882 | BC042788.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P500882 | BC115364.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P500882 | AJ223765.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P500882 | AK079401.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P500882 | AK172251.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P500882 | BC042788.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112896 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P140067 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06473141 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06473151 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06473143 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06473153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948534 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24948536 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24948524 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24948534 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24948547 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06213562 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05905913 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24948538 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24948528 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06473148 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24948522 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24948516 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24948530 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24948532 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24948541 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24948537 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24948545 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06580369 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05933065 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24948544 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24948518 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24948527 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10549650 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549651 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549652 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549649 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549648 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422089_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10549647 | - | - |
| Mouse Genome 430 2.0 Array | 1422089_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1422089_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17086_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17086_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700000410.mm.1 | NM_010746.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000410.mm.1 | NM_010746.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000410.mm.1 | TC0700000032.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700000413.mm.1 | NM_010746.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000413.mm.1 | NM_010746.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000413.mm.1 | TC0700000032.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422089_at | - | - |
| Mouse Expression Set 430 | 1422089_PM_at | - | - |
| Murine Genome U74v2 Set | 97781_at | - | - |
| Murine Genome U74v2 Set | 97781_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422089_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422089_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422089_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422089_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 845230 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2450026 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002450026 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2747294 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2747294 | - | - |
| mouse-6 v1.1 (Illumina) | scl33136.6.1_30-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2450026 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl33136.6.1_30-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2450026 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2747294 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2747294 | - | - |

Ncr1
Genomic Map