Assay Details
| Gene Symbol: Peg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008817.2 | NP_032843.2 | 1 - 2 | 161 | 125 | |
| GenBank mRNA | AK044602.1 | - | 1 - 2 | 162 | 125 | |
| AK053523.1 | - | 1 - 2 | 162 | 125 | ||
| AK122238.1 | - | 1 - 2 | 140 | 125 | ||
| AK141217.1 | - | 1 - 2 | 162 | 125 | ||
| AK163845.1 | - | 1 - 2 | 164 | 125 | ||
| BC072661.1 | - | 1 - 2 | 149 | 30543809 | 125 | |
| BC085183.1 | - | 1 - 2 | 146 | 30663888 | 125 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK141217.1, AK163845.1, BC072661.1, BC085183.1, NM_008817.2 transcripts.
Set Membership: |
|

Peg3
Genomic Map