Assay Details
| Gene Symbol: Peg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008817.2 | NP_032843.2 | 5 - 6 | 551 | 95 | |
| GenBank mRNA | AB003040.1 | - | 5 - 6 | 548 | 95 | |
| AF038939.1 | - | 2 - 3 | 202 | 95 | ||
| AK044602.1 | - | 5 - 6 | 552 | 95 | ||
| AK122238.1 | - | 5 - 6 | 530 | 95 | ||
| AK141217.1 | - | 5 - 6 | 552 | 95 | ||
| AK163845.1 | - | 5 - 6 | 554 | 95 | ||
| BC072661.1 | - | 5 - 6 | 539 | 30543809 | 95 | |
| BC085183.1 | - | 5 - 6 | 536 | 30663888 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Peg3
Genomic Map