Assay Details
| Gene Symbol: Peg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008817.2 | NP_032843.2 | 6 - 7 | 655 | 79 | |
| GenBank mRNA | AB003040.1 | - | 6 - 7 | 652 | 79 | |
| AF038939.1 | - | 3 - 4 | 306 | 79 | ||
| AK122238.1 | - | 6 - 7 | 634 | 79 | ||
| AK141217.1 | - | 6 - 7 | 656 | 79 | ||
| AK163845.1 | - | 6 - 7 | 658 | 79 | ||
| BC072661.1 | - | 6 - 7 | 643 | 30543809 | 79 | |
| BC085183.1 | - | 6 - 7 | 640 | 30663888 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Peg3
Genomic Map