Assay Details
| Gene Symbol: Trpc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009428.2 | NP_033454.1 | 3 - 4 | 1310 | 69 | |
| GenBank mRNA | AF060107.1 | - | 3 - 4 | 1309 | 69 | |
| AJ006204.1 | - | 2 - 3 | 907 | 69 | ||
| AK038531.1 | - | 3 - 4 | 1134 | 69 | ||
| BC112972.1 | - | 3 - 4 | 1128 | 40049027 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trpc5
Genomic Map