Assay Details
| Gene Symbol: Trpc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009428.2 | NP_033454.1 | 5 - 6 | 1773 | 71 | |
| GenBank mRNA | AF029983.1 | - | 5 - 6 | 1428 | 71 | |
| AF060107.1 | - | 5 - 6 | 1772 | 71 | ||
| AJ006204.1 | - | 4 - 5 | 1370 | 71 | ||
| AK038531.1 | - | 5 - 6 | 1597 | 71 | ||
| AK047867.1 | - | 3 - 4 | 794 | 71 | ||
| BC112972.1 | - | 5 - 6 | 1591 | 40049027 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trpc5
Genomic Map