Assay Details
| Gene Symbol: Trpc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009428.2 | NP_033454.1 | 9 - 10 | 2546 | 77 | |
| GenBank mRNA | AF029983.1 | - | 9 - 10 | 2201 | 77 | |
| AF060107.1 | - | 9 - 10 | 2545 | 77 | ||
| AJ006204.1 | - | 8 - 9 | 2143 | 77 | ||
| AK047867.1 | - | 7 - 8 | 1567 | 77 | ||
| BC112972.1 | - | 9 - 10 | 2364 | 40049027 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trpc5
Genomic Map