Assay Details
Gene Symbol: Nostrin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181547.3 | NP_853525.3 | 13 - 14 | 1272 | 66 | |
GenBank mRNA | AJ564264.1 | - | 13 - 14 | 1182 | 66 | |
AY280963.1 | - | 13 - 14 | 1225 | 66 | ||
BC069942.1 | - | 13 - 14 | 1249 | 30443942 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
