Assay Details
Gene Symbol: Nostrin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181547.3 | NP_853525.3 | 14 - 15 | 1383 | 125 | |
GenBank mRNA | AJ564264.1 | - | 14 - 15 | 1293 | 125 | |
AY280963.1 | - | 14 - 15 | 1336 | 125 | ||
BC069942.1 | - | 14 - 15 | 1360 | 30443942 | 125 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
