Assay Details
| Gene Symbol: Olfr52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146583.1 | NP_666794.1 | 1 | 541 | 110 | |
| GenBank mRNA | AF102528.1 | - | 1 | 352 | 110 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2094781 | NM_146583.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 508474 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P148457 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2094781 | NM_146583.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P310109 | AF102528.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P310109 | NM_146583.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P148457 | - | - |
| Whole Mouse Genome G4122A | A_51_P310109 | AF102528.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P310109 | NM_146583.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131693 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P148457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401041 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01745304 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401102 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401091 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401093 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401104 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401108 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401138 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401119 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401112 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401096 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401124 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401106 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401105 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401117 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401129 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401097 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401116 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401092 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401122 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401135 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01649443 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401133 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401099 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401090 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401098 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401125 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401127 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401111 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10484568 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10484567 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 327950 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3780253 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003780253 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2688543 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2688543 | - | - |
| mouse-6 v1.1 (Illumina) | scl19053.1.1_1-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3780253 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl19053.1.1_1-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3780253 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2688543 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2688543 | - | - |

Olfr52
Genomic Map