Assay Details
| Gene Symbol: Chrm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033269.4 | NP_150372.1 | 1 - 2 | 535 | 102 | |
| GenBank mRNA | AK051841.1 | - | 1 - 2 | 580 | 102 | |
| AK080950.1 | - | 1 - 2 | 536 | 102 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK080950.1, NM_033269.4 transcripts.
Set Membership: |
|

Chrm3
Genomic Map