Assay Details
| Gene Symbol: Chrm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033269.4 | NP_150372.1 | 2 - 3 | 654 | 66 | |
| GenBank mRNA | AK051841.1 | - | 2 - 3 | 699 | 66 | |
| BC129892.1 | - | 2 - 3 | 149 | 40111498 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC129892.1, NM_033269.4 transcripts.
Set Membership: |
|

Chrm3
Genomic Map