Assay Details
| Gene Symbol: Olfr1346 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146916.1 | NP_667127.1 | 1 | 662 | 84 | |
| GenBank mRNA | BC137876.1 | - | 1 | 686 | 8860895 | 84 |
| BC137877.1 | - | 1 | 686 | 8860896 | 84 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P514601 | NM_146916.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P514601 | BC137876.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P514601 | BC137877.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P514601 | NM_146916.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P514601 | BC137876.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P514601 | BC137877.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 904961 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P124277 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P514601 | NM_146916.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P514601 | BC137876.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P514601 | BC137877.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P514601 | NM_146916.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P514601 | BC137876.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P514601 | BC137877.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P514601 | NM_146916.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P514601 | BC137876.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P514601 | BC137877.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P514601 | NM_146916.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P514601 | BC137876.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P514601 | BC137877.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955620 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955611 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955642 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955647 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955640 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955637 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955633 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955618 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955644 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955625 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955629 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955635 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955612 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955616 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955626 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955624 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955610 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955614 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955621 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955643 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955648 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955630 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955628 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955638 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955623 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955613 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955634 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955646 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P124277 | - | - |
| Whole Mouse Genome G4122A | A_51_P514601 | NM_146916.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P514601 | BC137876.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P514601 | BC137877.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P514601 | NM_146916.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P514601 | BC137876.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P514601 | BC137877.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P140155 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P124277 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06016407 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06205243 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06016403 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06016404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955626 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955608 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955621 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955597 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955623 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955603 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955607 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955651 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955645 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955601 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955633 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06016407 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955620 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955646 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06016395 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06621026 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955612 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955644 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955613 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06016399 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955648 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05890547 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955591 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955614 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955581 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05941939 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05943571 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06016403 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955642 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955604 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955629 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06016402 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05943564 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955647 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955592 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955583 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06016401 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955630 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955625 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955656 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955649 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955611 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955585 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06607424 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955599 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05943572 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955586 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955618 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955610 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955617 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955655 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06010866 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955626 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10549872 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10549871 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700000900.mm.1 | TC0700000082.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 477623 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 580451 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 580451 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000580451 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1247690 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1247690 | - | - |
| mouse-6 v1.1 (Illumina) | scl9729.1.1_68-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 580451 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl9729.1.1_68-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 580451 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1247690 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1247690 | - | - |

Olfr1346
Genomic Map