Assay Details
| Gene Symbol: Ushbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181418.3 | NP_852083.2 | 2 - 3 | 218 | 129 | |
| GenBank mRNA | AK043798.1 | - | 2 - 3 | 129 | ||
| BC026453.1 | - | 2 - 3 | 202 | 4460920 | 129 | |
| BY241807.1 | - | 2 - 3 | 234 | 129 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC026453.1,NM_181418.3 transcripts.
Set Membership: |
|

Ushbp1
Genomic Map