Assay Details
| Gene Symbol: Ushbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181418.3 | NP_852083.2 | 9 - 10 | 1593 | 64 | |
| GenBank mRNA | AK043798.1 | - | 9 - 10 | 1531 | 64 | |
| AK084049.1 | - | 6 - 7 | 1040 | 64 | ||
More Information
Set Membership: |
|

Ushbp1
Genomic Map