Assay Details
| Gene Symbol: Mvb12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028617.2 | NP_082893.1 | 2 - 3 | 226 | 85 | |
| GenBank mRNA | AK003644.1 | - | 2 - 3 | 206 | 85 | |
| BC020119.1 | - | 2 - 3 | 226 | 3982577 | 85 | |
| BC027419.1 | - | 2 - 3 | 202 | 4163554 | 85 | |
| BC087734.1 | - | 2 - 3 | 223 | 5101734 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mvb12a
Genomic Map