Assay Details
| Gene Symbol: Mvb12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028617.2 | NP_082893.1 | 4 - 5 | 449 | 70 | |
| GenBank mRNA | AK003644.1 | - | 4 - 5 | 429 | 70 | |
| AK199858.1 | - | 1 - 2 | 49 | 70 | ||
| BC020119.1 | - | 4 - 5 | 449 | 3982577 | 70 | |
| BC027419.1 | - | 4 - 5 | 425 | 4163554 | 70 | |
| BC087734.1 | - | 4 - 5 | 446 | 5101734 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mvb12a
Genomic Map