Assay Details
| Gene Symbol: Mvb12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028617.2 | NP_082893.1 | 5 - 6 | 566 | 125 | |
| GenBank mRNA | AK003644.1 | - | 5 - 6 | 546 | 125 | |
| BC020119.1 | - | 5 - 6 | 566 | 3982577 | 125 | |
| BC027419.1 | - | 5 - 6 | 542 | 4163554 | 125 | |
| BC087734.1 | - | 5 - 6 | 563 | 5101734 | 125 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mvb12a
Genomic Map