Assay Details
| Gene Symbol: Tmem14c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025387.3 | NP_079663.1 | 1 - 2 | 148 | 99 | |
| GenBank mRNA | AK003891.1 | - | 1 - 2 | 97 | 99 | |
| AK008878.1 | - | 1 - 2 | 70 | 99 | ||
| AK010609.1 | - | 1 - 2 | 108 | 99 | ||
| AK012101.1 | - | 1 - 2 | 95 | 99 | ||
| AK019395.1 | - | 1 - 2 | 97 | 99 | ||
| BC055862.1 | - | 1 - 2 | 72 | 3492090 | 93 | |
| BY785978.1 | - | 1 - 2 | 148 | 99 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK003891.1, AK008878.1, AK010609.1, AK012101.1, AK019395.1, BC055862.1, NM_025387.3 transcripts.
Set Membership: |
|

Tmem14c
Genomic Map