Assay Details
| Gene Symbol: Tmem14c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025387.3 | NP_079663.1 | 4 - 5 | 381 | 65 | |
| GenBank mRNA | AK003891.1 | - | 4 - 5 | 330 | 65 | |
| AK008495.1 | - | 3 - 4 | 271 | 65 | ||
| AK008878.1 | - | 4 - 5 | 303 | 65 | ||
| AK010609.1 | - | 4 - 5 | 341 | 65 | ||
| AK012101.1 | - | 4 - 5 | 328 | 65 | ||
| AK019395.1 | - | 4 - 5 | 330 | 65 | ||
| AK050688.1 | - | 4 - 5 | 316 | 65 | ||
| BC025854.1 | - | 4 - 5 | 282 | 5250659 | 65 | |
| BC069899.1 | - | 3 - 4 | 1027 | 65 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tmem14c
Genomic Map