Assay Details
| Gene Symbol: Tnmd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022322.2 | NP_071717.1 | 1 - 2 | 251 | 100 | |
| GenBank mRNA | AB055422.1 | - | 1 - 2 | 58 | 100 | |
| AF191768.1 | - | 1 - 2 | 197 | 100 | ||
| AF219993.1 | - | 1 - 2 | 124 | 100 | ||
| AF291655.1 | - | 1 - 2 | 276 | 100 | ||
| AK003748.1 | - | 1 - 2 | 137 | 100 | ||
| AK014761.1 | - | 1 - 2 | 100 | |||
| BC006919.1 | - | 1 - 2 | 60 | 3601223 | 100 | |
| BC049944.1 | - | 1 - 2 | 251 | 2811628 | 100 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK014761.1 transcripts.
Set Membership: |
|

Tnmd
Genomic Map