Assay Details
| Gene Symbol: Tnmd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022322.2 | NP_071717.1 | 4 - 5 | 621 | 142 | |
| GenBank mRNA | AB055422.1 | - | 4 - 5 | 428 | 142 | |
| AF191768.1 | - | 4 - 5 | 567 | 142 | ||
| AF219993.1 | - | 4 - 5 | 494 | 142 | ||
| AF291655.1 | - | 4 - 5 | 646 | 142 | ||
| AK003748.1 | - | 4 - 5 | 140 | |||
| AK014761.1 | - | 4 - 5 | 460 | 142 | ||
| BC006919.1 | - | 4 - 5 | 430 | 3601223 | 142 | |
| BC049944.1 | - | 4 - 5 | 621 | 2811628 | 142 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK014761.1 transcripts.
Set Membership: |
|

Tnmd
Genomic Map