Assay Details
| Gene Symbol: Kbtbd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145958.2 | NP_666070.2 | 3 - 4 | 985 | 109 | |
| GenBank mRNA | AK028396.1 | - | 3 - 4 | 931 | 109 | |
| AK129374.1 | - | 3 - 4 | 895 | 109 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kbtbd2
Genomic Map