Assay Details
Gene Symbol: Mctp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030174.2 | NP_084450.2 | 11 - 12 | 1459 | 128 | |
GenBank mRNA | AK047562.1 | - | 11 - 12 | 1460 | 128 | |
AK054478.1 | - | 7 - 8 | 832 | 128 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
