Assay Details
| Gene Symbol: Fgfr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008011.2 | NP_032037.2 | 1 - 2 | 108 | 79 | |
| GenBank mRNA | AK084850.1 | - | 1 - 2 | 108 | 79 | |
| AK085723.1 | - | 1 - 2 | 107 | 79 | ||
| BC033313.1 | - | 1 - 2 | 295 | 5132831 | 79 | |
| X59927.1 | - | 1 - 2 | 51 | 79 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK084850.1, BC033313.1, NM_008011.2, X59927.1 transcripts.
Set Membership: |
|

Fgfr4
Genomic Map