Assay Details
| Gene Symbol: Fgfr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008011.2 | NP_032037.2 | 7 - 8 | 1073 | 64 | |
| GenBank mRNA | AK084850.1 | - | 7 - 8 | 1073 | 64 | |
| AK085723.1 | - | 7 - 8 | 1071 | 64 | ||
| AY493377.2 | - | 6 - 7 | 959 | 64 | ||
| BC033313.1 | - | 7 - 8 | 1260 | 5132831 | 64 | |
| DQ388428.1 | - | 6 - 7 | 959 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fgfr4
Genomic Map