Assay Details
| Gene Symbol: Nubp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011955.2 | NP_036085.1 | 6 - 7 | 494 | 87 | |
| GenBank mRNA | AF114170.1 | - | 6 - 7 | 471 | 87 | |
| AK157212.1 | - | 6 - 7 | 486 | 87 | ||
| AK214196.1 | - | 1 - 2 | 93 | 87 | ||
| AK218168.1 | - | 1 - 2 | 93 | 87 | ||
| BC055436.1 | - | 6 - 7 | 482 | 3660324 | 87 | |
| CX210766.1 | - | 3 - 4 | 182 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nubp1
Genomic Map