Assay Details
Gene Symbol: Crebbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025432.1 | NP_001020603.1 | 11 - 12 | 2336 | 66 | |
GenBank mRNA | BC072594.1 | - | 11 - 12 | 2322 | 66 | |
S66385.1 | - | 11 - 12 | 2155 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
