Assay Details
| Gene Symbol: Slx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177472.5 | NP_803423.2 | 4 - 5 | 938 | 76 | |
| GenBank mRNA | BC065125.1 | - | 4 - 5 | 943 | 30534878 | 76 |
More Information
Updated Mapping Information
Set Membership: |
|

Slx4
Genomic Map