Assay Details
| Gene Symbol: Slx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177472.5 | NP_803423.2 | 5 - 6 | 1146 | 116 | |
| GenBank mRNA | AK173263.1 | - | 4 - 5 | 559 | 116 | |
| BC065125.1 | - | 5 - 6 | 1151 | 30534878 | 116 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slx4
Genomic Map