Assay Details
| Gene Symbol: Tmem44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172614.3 | NP_766202.3 | 3 - 4 | 439 | 80 | |
| GenBank mRNA | AK031524.1 | - | 3 - 4 | 401 | 80 | |
| AK045675.1 | - | 4 - 5 | 502 | 80 | ||
| AK048655.1 | - | 3 - 4 | 440 | 80 | ||
| AK082652.1 | - | 3 - 4 | 1674 | 80 | ||
| BC059216.1 | - | 3 - 4 | 777 | 80 | ||
| BC080307.1 | - | 4 - 5 | 335 | 80 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK045675.1, AK048655.1 transcripts.
Set Membership: |
|

Tmem44
Genomic Map