Assay Details
| Gene Symbol: Tmem44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK045675.1 | - | 10 - 11 | 1336 | 97 | |
| BC080307.1 | - | 10 - 11 | 1169 | 97 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem44| Gene Symbol: Tmem44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK045675.1 | - | 10 - 11 | 1336 | 97 | |
| BC080307.1 | - | 10 - 11 | 1169 | 97 | ||
Set Membership: |
|