Assay Details
| Gene Symbol: Pkp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026163.2 | NP_080439.1 | 10 - 11 | 2066 | 99 | |
| GenBank mRNA | AK004650.1 | - | 10 - 11 | 2067 | 99 | |
| AK005020.1 | - | 10 - 11 | 2098 | 99 | ||
| AK049481.1 | - | 10 - 11 | 2067 | 99 | ||
| AK167660.1 | - | 10 - 11 | 2068 | 99 | ||
| BC018487.1 | - | 4 - 5 | 622 | 99 | ||
| BC037133.1 | - | 5 - 6 | 672 | 99 | ||
| BC068279.1 | - | 8 - 9 | 1347 | 99 | ||
| BC138252.1 | - | 10 - 11 | 2066 | 8861271 | 99 | |
| BC138253.1 | - | 10 - 11 | 2066 | 8861272 | 99 | |
More Information
Set Membership: |
|

Pkp2
Genomic Map