Assay Details
| Gene Symbol: Pkp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026163.2 | NP_080439.1 | 12 - 13 | 2342 | 69 | |
| GenBank mRNA | AK004650.1 | - | 12 - 13 | 2343 | 69 | |
| AK005020.1 | - | 12 - 13 | 2374 | 69 | ||
| AK049481.1 | - | 12 - 13 | 2343 | 69 | ||
| AK167660.1 | - | 12 - 13 | 2344 | 69 | ||
| BC018487.1 | - | 6 - 7 | 898 | 69 | ||
| BC037133.1 | - | 7 - 8 | 948 | 69 | ||
| BC068279.1 | - | 10 - 11 | 1623 | 69 | ||
| BC138252.1 | - | 12 - 13 | 2342 | 8861271 | 69 | |
| BC138253.1 | - | 12 - 13 | 2342 | 8861272 | 69 | |
More Information
Set Membership: |
|

Pkp2
Genomic Map