Assay Details
| Gene Symbol: Pkp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026163.2 | NP_080439.1 | 1 - 2 | 251 | 101 | |
| GenBank mRNA | AK004650.1 | - | 1 - 2 | 252 | 101 | |
| AK004731.2 | - | 1 - 2 | 251 | 101 | ||
| AK005020.1 | - | 1 - 2 | 283 | 101 | ||
| AK049481.1 | - | 1 - 2 | 252 | 101 | ||
| AK162000.1 | - | 1 - 2 | 252 | 101 | ||
| AK167660.1 | - | 1 - 2 | 253 | 101 | ||
| BC138252.1 | - | 1 - 2 | 251 | 8861271 | 101 | |
| BC138253.1 | - | 1 - 2 | 251 | 8861272 | 101 | |
More Information
Set Membership: |
|

Pkp2
Genomic Map