Assay Details
| Gene Symbol: Pkp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026163.2 | NP_080439.1 | 6 - 7 | 1456 | 106 | |
| GenBank mRNA | AK004650.1 | - | 6 - 7 | 1457 | 106 | |
| AK005020.1 | - | 6 - 7 | 1488 | 106 | ||
| AK049481.1 | - | 6 - 7 | 1457 | 106 | ||
| AK167660.1 | - | 6 - 7 | 1458 | 106 | ||
| BC037133.1 | - | 1 - 2 | 62 | 106 | ||
| BC068279.1 | - | 4 - 5 | 736 | 106 | ||
| BC138252.1 | - | 6 - 7 | 1456 | 8861271 | 106 | |
| BC138253.1 | - | 6 - 7 | 1456 | 8861272 | 106 | |
More Information
Set Membership: |
|

Pkp2
Genomic Map